1-(2-Chlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
1-[(2-Chlorophenyl)methyl]-2-oxo-1,2-dihydropyridine-3-carboxylic acid
043436-1G
043436-5G
043436-500MG
101860-786EA
806.15
USD
101860-786
101860-782
101860-784
1-(2-Chlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid ≥95%
1-(2-Chlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid
Hazard: Irritant
Formula:
C₁₃H₁₀ClNO₃ MW: 263.68 g/mol Melting Pt: 170…174 °C |
MDL Number:
MFCD00140815 CAS Number: 66158-19-8 |
Specification Test Results
Melting point | 170-174°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...