3-(Dimethylamino)-2-(4-fluorophenyl)acrylonitrile ≥95%

Supplier: Matrix Scientific

043419-500MG 043419-5G 043419-1G
101860-688EA 181.54 USD
101860-688 101860-690 101860-692
3-(Dimethylamino)-2-(4-fluorophenyl)acrylonitrile ≥95%
3-(Dimethylamino)-2-(4-fluorophenyl)acrylonitrile

Hazard: Irritant

Formula: C₁₁H₁₁FN₂
Melting Pt: 71…73 °C
MDL Number: MFCD00138582
CAS Number: 320416-88-4

Order Now

Specification Test Results

Melting point 71-73°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR