6-Phenyl[1,2,4]triazolo[1,5-a]pyrimidin-7-ylamine ≥95%

Supplier: Matrix Scientific

043414-5G 043414-1G 043414-500MG
101860-658EA 375.38 USD
101860-658 101860-660 101860-662
6-Phenyl[1,2,4]triazolo[1,5-a]pyrimidin-7-ylamine ≥95%
6-Phenyl[1,2,4]triazolo[1,5-a]pyrimidin-7-ylamine

Hazard: Irritant

Formula: C₁₁H₉N₅
Melting Pt: 282…284 °C
MDL Number: MFCD00138731
CAS Number: 320416-33-9

Order Now

Specification Test Results

Melting point 282-284°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR