5-Acetyl-2,2-dimethyl-1,3-dioxane-4,6-dione (5-acetylmeldrum's acid) ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Acetyl-2,2-dimethyl-1,3-dioxane-4,6-dione

043412-500MG 043412-5G 043412-1G
101860-646EA 149.5 USD
101860-646 101860-648 101860-650
5-Acetyl-2,2-dimethyl-1,3-dioxane-4,6-dione (5-acetylmeldrum's acid) ≥95%
5-Acetyl-2,2-dimethyl-1,3-dioxane-4,6-dione (5-Acetylmeldrum's acid)

Hazard: Irritant

Order Now

Specification Test Results

Melting point 83-85°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR