1-[2-Amino-5-(4-methylpiperidino)phenyl]-1-ethanone ≥95%
Supplier: Matrix Scientific
043399-1G
043399-5G
043399-500MG
101860-590EA
376.92
USD
101860-590
101860-592
101860-594
1-[2-Amino-5-(4-methylpiperidino)phenyl]-1-ethanone ≥95%
1-[2-Amino-5-(4-methylpiperidino)phenyl]-1-ethanone
Hazard: Irritant
Formula:
C₁₄H₂₀N₂O MW: 232.33 g/mol Melting Pt: 79…81 °C |
MDL Number:
MFCD08276955 CAS Number: 886361-39-3 |
Specification Test Results
Melting point | 79-81°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...