6-(4-Formylphenyl)nicotinonitrile ≥95%
Supplier: Matrix Scientific
043389-5G
043389-1G
043389-500MG
101860-538EA
278.46
USD
101860-538
101860-540
101860-542
6-(4-Formylphenyl)nicotinonitrile ≥95%
6-(4-Formylphenyl)nicotinonitrile
Hazard: Irritant
Formula:
C₁₃H₈N₂O Melting Pt: 200…202 °C |
MDL Number:
MFCD08276960 CAS Number: 851340-81-3 |
Specification Test Results
Melting point | 200-202°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...