4-(5-Butyl-1,3,4-oxadiazol-2-yl)phenylamine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-[5-(But-1-yl)-1,3,4-oxadiazol-2-yl]aniline

043380-1G 043380-5G 043380-500MG
101860-500EA 373.75 USD
101860-500 101860-502 101860-504
4-(5-Butyl-1,3,4-oxadiazol-2-yl)phenylamine ≥95%
4-(5-Butyl-1,3,4-oxadiazol-2-yl)phenylamine

Hazard: Irritant

Formula: C₁₂H₁₅N₃O
MW: 217.27 g/mol
Melting Pt: 119…121 °C
MDL Number: MFCD06797387
CAS Number: 100933-82-2

Order Now

Specification Test Results

Melting point 119-121°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR