2-Hydroxy-4-methoxybenzenecarbohydrazide ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Hydroxy-4-methoxybenzohydrazide
043378-1G
043378-5G
043378-500MG
101860-490EA
232.31
USD
101860-490
101860-492
101860-488
2-Hydroxy-4-methoxybenzenecarbohydrazide ≥95%
2-Hydroxy-4-methoxybenzenecarbohydrazide
Hazard: Irritant
Formula:
C₈H₁₀N₂O₃ MW: 182.18 g/mol Melting Pt: 173…175 °C |
MDL Number:
MFCD06797386 CAS Number: 41697-08-9 |
Specification Test Results
Melting point | 173-175°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...