6-Chloroimidazo[1,2-a]pyridine-2-carbohydrazide ≥95%
Supplier: Matrix Scientific
043376-1G
043376-5G
043376-500MG
101860-478EA
556.92
USD
101860-478
101860-480
101860-482
6-Chloroimidazo[1,2-a]pyridine-2-carbohydrazide ≥95%
6-Chloroimidazo[1,2-a]pyridine-2-carbohydrazide
Hazard: Irritant
Formula:
C₈H₇ClN₄O Melting Pt: 235…237 °C |
MDL Number:
MFCD06797385 CAS Number: 119448-28-1 |
Specification Test Results
Melting point | 235-237°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...