4-(3,4-Dimethoxyphenyl)-5-methyl-2,4-dihydro-3H-1,2,4-triazol-3-one ≥95%

Supplier: Matrix Scientific

043350-5G 043350-500MG 043350-1G
101860-380EA 278.46 USD
101860-380 101860-382 101860-384
4-(3,4-Dimethoxyphenyl)-5-methyl-2,4-dihydro-3H-1,2,4-triazol-3-one ≥95%
4-(3,4-Dimethoxyphenyl)-5-methyl-2,4-dihydro-3H-1,2,4-triazol-3-one

Hazard: Irritant

Formula: C₁₁H₁₃N₃O₃
MW: 235.24 g/mol
Melting Pt: 222…224 °C
MDL Number: MFCD06496114
CAS Number: 860787-33-3

Order Now

Specification Test Results

Melting point 222-224°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR