Ethyl-6,8-dibromo-2-oxo-1,2-dihydro-3-quinolinecarboxylate ≥95%

Supplier: Matrix Scientific

043334-500MG 043334-1G 043334-5G
101860-292EA 278.46 USD
101860-292 101860-294 101860-296
Ethyl-6,8-dibromo-2-oxo-1,2-dihydro-3-quinolinecarboxylate ≥95%
Ethyl-6,8-dibromo-2-oxo-1,2-dihydro-3-quinolinecarboxylate

Hazard: Irritant

Formula: C₁₂H₉Br₂NO₃
MW: 375.03 g/mol
Melting Pt: 233…235 °C
MDL Number: MFCD04124609
CAS Number: 860787-14-0

Order Now

Specification Test Results

Melting point 233-235°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR