5-Methyl-4-phenyl-2,4-dihydro-3H-1,2,4-triazol-3-one ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Methyl-4-phenyl-1H-1,2,4-triazol-5(4H)-one

043315-1G 043315-5G 043315-500MG
101860-190EA 215.38 USD
101860-190 101860-192 101860-188
5-Methyl-4-phenyl-2,4-dihydro-3H-1,2,4-triazol-3-one ≥95%
5-Methyl-4-phenyl-2,4-dihydro-3H-1,2,4-triazol-3-one

Hazard: Irritant

Formula: C₉H₉N₃O
MW: 175.19 g/mol
Melting Pt: 153…154 °C
MDL Number: MFCD03787260
CAS Number: 1010-54-4

Order Now

Specification Test Results

Melting point 153-154°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR