Methyl-7-amino-2-methylpyrazolo[1,5-a]pyrimidine-6-carboxylate ≥95%

Supplier: Matrix Scientific

043293-1G 043293-500MG 043293-5G
101860-084EA 280.6 USD
101860-084 101860-086 101860-088
Methyl-7-amino-2-methylpyrazolo[1,5-a]pyrimidine-6-carboxylate ≥95%
Methyl-7-amino-2-methylpyrazolo[1,5-a]pyrimidine-6-carboxylate

Hazard: Irritant

Formula: C₉H₁₀N₄O₂
Melting Pt: 186…188 °C
MDL Number: MFCD03102683
CAS Number: 691869-96-2

Order Now

Specification Test Results

Melting point 186-188°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR