4,5-Dihydroxy-1-methyltetrahydro-2H-imidazole-2-thione ≥95%

Supplier: Matrix Scientific

043290-500MG
101860-074EA 208.15 USD
101860-074
4,5-Dihydroxy-1-methyltetrahydro-2H-imidazole-2-thione ≥95%
4,5-Dihydroxy-1-methyltetrahydro-2H-imidazole-2-thione

Hazard: Irritant

Formula: C₄H₈N₂O₂S
Melting Pt: 132…134 °C
MDL Number: MFCD03001193
CAS Number: 265986-74-1

Order Now

Specification Test Results

Melting point 132-134°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR