Methyl-1-(cyanomethyl)-4-piperidinecarboxylate ≥95%

Supplier: Matrix Scientific

043272-1G 043272-5G 043272-500MG
101860-008EA 375.38 USD
101860-008 101860-010 101860-012
Methyl-1-(cyanomethyl)-4-piperidinecarboxylate ≥95%
Methyl-1-(cyanomethyl)-4-piperidinecarboxylate

Hazard: Irritant

Formula: C₉H₁₄N₂O₂
MW: 182.22 g/mol
Melting Pt: 48…50 °C
MDL Number: MFCD01568888
CAS Number: 320424-43-9

Order Now

Specification Test Results

Melting point 48-50°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR