5-Amino-2-[(4-methoxyphenyl)sulfanyl]benzenecarbonitrile ≥95%

Supplier: Matrix Scientific

043231-500MG 043231-5G 043231-1G
101859-810EA 1073.85 USD
101859-810 101859-806 101859-808
5-Amino-2-[(4-methoxyphenyl)sulfanyl]benzenecarbonitrile ≥95%
5-Amino-2-[(4-methoxyphenyl)sulfanyl]benzenecarbonitrile

Hazard: Irritant

Formula: C₁₄H₁₂N₂OS
MW: 256.33 g/mol
Melting Pt: 120…121 °C
MDL Number: MFCD00793542
CAS Number: 306980-92-7

Order Now

Specification Test Results

Melting point 120-121°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR