5-Methyl-1-(4-nitrophenyl)-1H-1,2,4-triazole-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

043215-500MG 043215-1G
101859-730EA 1061.54 USD
101859-730 101859-728
5-Methyl-1-(4-nitrophenyl)-1H-1,2,4-triazole-3-carboxylic acid ≥95%
5-Methyl-1-(4-nitrophenyl)-1H-1,2,4-triazole-3-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₈N₄O₄
Melting Pt: 203…205 °C
MDL Number: MFCD00173063
CAS Number: 1025-88-3

Order Now

Specification Test Results

Melting point 203-205°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR