Ethyl-2-(4-chlorophenyl)-1,3-thiazole-4-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 2-(4-chlorophenyl)thiazole-4-carboxylate
043212-5G
043212-1G
043212-500MG
101859-710EA
181.54
USD
101859-710
101859-714
101859-712
Ethyl-2-(4-chlorophenyl)-1,3-thiazole-4-carboxylate ≥95%
Ethyl-2-(4-chlorophenyl)-1,3-thiazole-4-carboxylate
Hazard: Irritant
Formula:
C₁₂H₁₀ClNO₂S MW: 267.74 g/mol Melting Pt: 99…101 °C |
MDL Number:
MFCD00142031 CAS Number: 61786-00-3 |
Specification Test Results
Melting point | 99-101°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...