Methyl-1-(4-chlorophenyl)-5-(2-methoxy-2-oxoethyl)1H-1,2,3-triazole-4-carboxylate ≥95%
Supplier: Matrix Scientific
043193-5G
043193-500MG
043193-1G
101859-604EA
467.69
USD
101859-604
101859-602
101859-606
Methyl-1-(4-chlorophenyl)-5-(2-methoxy-2-oxoethyl)1H-1,2,3-triazole-4-carboxylate ≥95%
Methyl-1-(4-chlorophenyl)-5-(2-methoxy-2-oxoethyl)1H-1,2,3-triazole-4-carboxylate
Hazard: Irritant
Formula:
C₁₃H₁₂ClN₃O₄ Melting Pt: 124…126 °C |
MDL Number:
MFCD00100191 CAS Number: 114462-77-0 |
Specification Test Results
Melting point | 124-126°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...