2-Amino-4-(tert-butyl)-1,3-thiazole-5-carbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-4-(tert-butyl)thiazole-5-carbonitrile

043189-1G 043189-500MG
101859-588EA 698.46 USD
101859-588 101859-586
2-Amino-4-(tert-butyl)-1,3-thiazole-5-carbonitrile ≥95%
2-Amino-4-(tert-butyl)-1,3-thiazole-5-carbonitrile

Hazard: Irritant

Formula: C₈H₁₁N₃S
MW: 181.26 g/mol
Melting Pt: 189…191 °C
MDL Number: MFCD00214700
CAS Number: 303994-99-2

Order Now

Specification Test Results

Melting point 189-191°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR