3-[5-(Trifluoromethyl)-2-pyridinyl]aniline ≥95%

Supplier: Matrix Scientific

043178-5G 043178-1G 043178-500MG
101859-526EA 416.3 USD
101859-526 101859-528 101859-530
3-[5-(Trifluoromethyl)-2-pyridinyl]aniline ≥95%
3-[5-(Trifluoromethyl)-2-pyridinyl]aniline

Hazard: Irritant

Formula: C₁₂H₉F₃N₂
MW: 238.21 g/mol
Melting Pt: 84…86 °C
MDL Number: MFCD08276968
CAS Number: 886361-00-8

Order Now

Specification Test Results

Melting point 84-86°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR