Methyl-3-methyl-4-nitrobenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3-methyl-4-nitrobenzoate

043138-1G
101859-362EA 232.31 USD
101859-362
Methyl-3-methyl-4-nitrobenzoate ≥95%
Methyl 3-methyl-4-nitrobenzoate

Hazard: Irritant

Formula: C₉H₉NO₄
MW: 195.18 g/mol
Melting Pt: 78…79 °C
MDL Number: MFCD00085640
CAS Number: 24078-21-5

Order Now

Specification Test Results

Melting point 82-83°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR