Pentafluorophenyl 4-fluorobenzenesulfonate ≥95%

Supplier: Matrix Scientific

043126-1G
101859-302EA 349.6 USD
101859-302
Pentafluorophenyl 4-fluorobenzenesulfonate ≥95%
Pentafluorophenyl 4-fluorobenzenesulphonate

Hazard: Irritant

Formula: C₁₂H₄F₆O₃S
Melting Pt: 46…48 °C
MDL Number: MFCD05975136
CAS Number: 885950-58-3

Order Now

Specification Test Results

Melting point 46-48°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR