5-(Methoxycarbonyl)-2-pyridine carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 5-(Methoxycarbonyl)picolinic acid

043124-5G
101859-290EA 33.35 USD
101859-290
5-(Methoxycarbonyl)-2-pyridine carboxylic acid ≥95%
5-(Methoxycarbonyl)-2-pyridine carboxylic acid

Hazard: Irritant

Formula: C₈H₇NO₄
MW: 181.15 g/mol
Melting Pt: 180…182 °C
MDL Number: MFCD00179343
CAS Number: 17874-79-2

Order Now

Specification Test Results

Melting point 180-182°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR