6-(1H-Pyrrol-1-yl)nicotinonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 6-(1H-Pyrrol-1-yl)pyridine-3-carbonitrile

043122-1G
101859-282EA 173.65 USD
101859-282
6-(1H-Pyrrol-1-yl)nicotinonitrile ≥95%
6-(1H-Pyrrol-1-yl)nicotinonitrile

Hazard: Irritant

Formula: C₁₀H₇N₃
MW: 169.19 g/mol
Melting Pt: 101…103 °C
MDL Number: MFCD00828638
CAS Number: 218157-81-4

Order Now

Specification Test Results

Melting point 101-103°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR