1-(4-Bromophenyl)-2,5-dimethyl-1H-pyrrole-3-carbaldehyde ≥95%

Supplier: Matrix Scientific

043120-1G
101859-272EA 349.6 USD
101859-272
1-(4-Bromophenyl)-2,5-dimethyl-1H-pyrrole-3-carbaldehyde ≥95%
1-(4-Bromophenyl)-2,5-dimethyl-1H-pyrrole-3-carbaldehyde

Hazard: Irritant

Formula: C₁₃H₁₂BrNO
Melting Pt: 117…119 °C
MDL Number: MFCD01009372
CAS Number: 347331-78-6

Order Now

Specification Test Results

Melting point 117-119°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR