2-(4-Chlorophenyl)-4-methyl-5-thiazolecarbohydrazide ≥95%
Supplier: Matrix Scientific
043116-500MG
043116-5G
101859-256EA
1600
USD
101859-256
101859-252
2-(4-Chlorophenyl)-4-methyl-5-thiazolecarbohydrazide ≥95%
2-(4-Chlorophenyl)-4-methyl-5-thiazolecarbohydrazide
Hazard: Irritant
Formula:
C₁₁H₈ClNO₂S Melting Pt: 234…236 °C |
MDL Number:
MFCD04112254 CAS Number: 685541-32-6 |
Specification Test Results
Melting point | 234-236°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...