1-(tert-Butyl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

043113-1G
101859-236EA 376.92 USD
101859-236
1-(tert-Butyl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid ≥95%
1-(tert-Butyl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid

Hazard: Irritant

Formula: C₉H₁₁F₃N₂O₂
Melting Pt: 115…117 °C
MDL Number: MFCD04124379
CAS Number: 142818-02-8

Order Now

Specification Test Results

Melting point 115-117°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR