2-(4-Hydroxy-1,3-thiazol-2-yl)ethanethioamide ≥95%

Supplier: Matrix Scientific

043111-1G
101859-224EA 232.31 USD
101859-224
2-(4-Hydroxy-1,3-thiazol-2-yl)ethanethioamide ≥95%
2-(4-Hydroxy-1,3-thiazol-2-yl)ethanethioamide

Hazard: Irritant

Formula: C₅H₆N₂OS₂
Melting Pt: >300 °C
MDL Number: MFCD04124364
CAS Number: 126106-28-3

Order Now

Specification Test Results

Melting point 300°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR