1-Ethyl-3-methyl-1H-pyrazol-5-amine ≥95%

Supplier: Matrix Scientific

043110-1G
101859-218EA 272.31 USD
101859-218
1-Ethyl-3-methyl-1H-pyrazol-5-amine ≥95%
1-Ethyl-3-methyl-1H-pyrazol-5-amine

Hazard: Irritant

Formula: C₆H₁₁N₃
Melting Pt: 99…100 °C
MDL Number: MFCD02255831
CAS Number: 3524-33-2

Order Now

Specification Test Results

Melting point 99-100°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR