5-(4-Methylphenyl)-1H-tetrazole ≥95%

Supplier: Matrix Scientific

Synonyms: 5-(p-Tolyl)-1H-tetrazole

043104-1G
101859-182EA 232.31 USD
101859-182
5-(4-Methylphenyl)-1H-tetrazole ≥95%
5-(4-Methylphenyl)-1H-tetrazole

Hazard: Irritant

Formula: C₈H₈N₄
MW: 160.18 g/mol
Melting Pt: 260 °C
MDL Number: MFCD01318164
CAS Number: 24994-04-5

Order Now

Specification Test Results

Melting point 260°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR