5-Bromo-7-nitro-1H-indazole ≥95%

Supplier: Matrix Scientific

043102-500MG
101859-170EA 280.6 USD
101859-170
5-Bromo-7-nitro-1H-indazole ≥95%
5-Bromo-7-nitro-1H-indazole

Hazard: Irritant

Formula: C₇H₄BrN₃O₂
Melting Pt: 220 °C
MDL Number: MFCD03617604
CAS Number: 316810-82-9

Order Now

Specification Test Results

Melting point 220°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR