7-Bromo-5-nitro-1H-indazole ≥95%
Supplier: Matrix Scientific
043101-1G
101859-166EA
236.92
USD
101859-166
7-Bromo-5-nitro-1H-indazole ≥95%
7-Bromo-5-nitro-1H-indazole
Hazard: Irritant
Formula:
C₇H₄BrN₃O₂ Melting Pt: 260…261 °C |
MDL Number:
MFCD03617603 CAS Number: 685109-10-8 |
Specification Test Results
Melting point | 260-261°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...