2-[2-(3-Furyl)vinyl]-5-hydroxy-1-benzofuran-3-carbaldehyde ≥95%

Supplier: Matrix Scientific

043099-1G
101859-154EA 434.7 USD
101859-154
2-[2-(3-Furyl)vinyl]-5-hydroxy-1-benzofuran-3-carbaldehyde ≥95%
2-[2-(3-Furyl)vinyl]-5-hydroxy-1-benzofuran-3-carbaldehyde

Hazard: Irritant

Formula: C₁₅H₁₀O₄
Melting Pt: 225…228 °C
MDL Number: MFCD03791159
CAS Number: 852691-02-2

Order Now

Specification Test Results

Melting point 225-228°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR