3-(4-Chlorostyryl)-1H-pyrazole ≥95%

Supplier: Matrix Scientific

043097-1G
101859-142EA 813.85 USD
101859-142
3-(4-Chlorostyryl)-1H-pyrazole ≥95%
3-(4-Chlorostyryl)-1H-pyrazole

Hazard: Irritant

Formula: C₁₁H₉ClN₂
Melting Pt: 138…140 °C
MDL Number: MFCD00120939
CAS Number: 852691-01-1

Order Now

Specification Test Results

Melting point 138-140°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR