3-(5-Chloro-1-benzofuran-2-yl)-1H-pyrazole ≥95%

Supplier: Matrix Scientific

043093-1G
101859-118EA 1400 USD
101859-118
3-(5-Chloro-1-benzofuran-2-yl)-1H-pyrazole ≥95%
3-(5-Chloro-1-benzofuran-2-yl)-1H-pyrazole

Hazard: Irritant

Formula: C₁₁H₇ClN₂O
Melting Pt: 195…197 °C
MDL Number: MFCD00112206
CAS Number: 852690-99-4

Order Now

Specification Test Results

Melting point 195-197°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR