1-(1,3-Benzodioxol-5-yl)-2-bromo-1-ethanone ≥95%

Supplier: Matrix Scientific

Synonyms: 5-(Bromoacetyl)-1,3-benzodioxole

043089-1G
101859-094EA 350.77 USD
101859-094
1-(1,3-Benzodioxol-5-yl)-2-bromo-1-ethanone ≥95%
1-(1,3-Benzodioxol-5-yl)-2-bromo-1-ethanone

Hazard: Irritant

Formula: C₉H₇BrO₃
MW: 243.06 g/mol
Melting Pt: 92…94 °C
MDL Number: MFCD01910077
CAS Number: 40288-65-1

Order Now

Specification Test Results

Melting point 92-94°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR