N'-Hydroxy-2-(methylsulfonyl)ethanimidamide ≥95%

Supplier: Matrix Scientific

043083-1G
101859-070EA 1163.08 USD
101859-070
N'-Hydroxy-2-(methylsulfonyl)ethanimidamide ≥95%
N'-Hydroxy-2-(methylsulfonyl)ethanimidamide

Hazard: Irritant

Formula: C₃H₈N₂O₃S
Melting Pt: 124…126 °C
MDL Number: MFCD00204089
CAS Number: 446276-20-6

Order Now

Specification Test Results

Melting point 124-126°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR