1,3-Dimethyl-1,6-dihydropyrazolo[3,4-c]pyrazole ≥95%

Supplier: Matrix Scientific

043081-1G
101859-058EA 927.69 USD
101859-058
1,3-Dimethyl-1,6-dihydropyrazolo[3,4-c]pyrazole ≥95%
1,3-Dimethyl-1,6-dihydropyrazolo[3,4-c]pyrazole

Hazard: Irritant

Formula: C₆H₈N₄
Melting Pt: 218…220 °C
MDL Number: MFCD00238783
CAS Number: 446276-19-3

Order Now

Specification Test Results

Melting point 218-220°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR