Ethyl-7-hydroxy-2-methyl-3-[3-(trifluoromethyl)phenyl]pyrazolo[1,5-a]pyrimidine-6-carboxylate ≥95%

Supplier: Matrix Scientific

043078-1G
101859-046EA 349.6 USD
101859-046
Ethyl-7-hydroxy-2-methyl-3-[3-(trifluoromethyl)phenyl]pyrazolo[1,5-a]pyrimidine-6-carboxylate ≥95%
Ethyl-7-hydroxy-2-methyl-3-[3-(trifluoromethyl)phenyl]pyrazolo[1,5-a]pyrimidine-6-carboxylate

Hazard: Irritant

Formula: C₁₇H₁₄F₃N₃O₃
MW: 365.31 g/mol
Melting Pt: 273…275 °C
MDL Number: MFCD03791151
CAS Number: 852690-95-0

Order Now

Specification Test Results

Melting point 273-275°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR