5-[(4-Nitrophenyl)methylene]-1,3-thiazolane-2,4-dione ≥95%

Supplier: Matrix Scientific

043074-1G
101859-022EA 173.65 USD
101859-022
5-[(4-Nitrophenyl)methylene]-1,3-thiazolane-2,4-dione ≥95%
5-[(4-Nitrophenyl)methylene]-1,3-thiazolane-2,4-dione

Hazard: Irritant

Formula: C₁₀H₆N₂O₄S
Melting Pt: 266…268 °C
MDL Number: MFCD00523219
CAS Number: 34301-40-1

Order Now

Specification Test Results

Melting point 266-268°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR