4-Chloro-6-methyl-2-(2-pyridinyl)pyrimidine ≥95%

Supplier: Matrix Scientific

043073-1G
101859-018EA 522.1 USD
101859-018
4-Chloro-6-methyl-2-(2-pyridinyl)pyrimidine ≥95%
4-Chloro-6-methyl-2-(2-pyridinyl)pyrimidine

Hazard: Irritant

Formula: C₁₀H₈ClN₃
MW: 205.65 g/mol
Melting Pt: 75…76 °C
MDL Number: MFCD00662786
CAS Number: 77168-31-1

Order Now

Specification Test Results

Melting point 75-76°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR