2-(2-Oxo-1,2-dihydro-3H-indol-3-yliden)-1-hydrazinecarbothioamide ≥95%

Supplier: Matrix Scientific

043072-5G 043072-500MG
101859-014EA 532.31 USD
101859-014 101859-010
2-(2-Oxo-1,2-dihydro-3H-indol-3-yliden)-1-hydrazinecarbothioamide ≥95%
2-(2-Oxo-1,2-dihydro-3H-indol-3-yliden)-1-hydrazinecarbothioamide

Hazard: Irritant

Formula: C₉H₈N₄OS
Melting Pt: 252…254 °C
MDL Number: MFCD00043581
CAS Number: 487-16-1

Order Now

Specification Test Results

Melting point 252-254°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR