4-Methoxy-3-(2-propynyloxy)benzenecarbaldehydeoxime ≥95%

Supplier: Matrix Scientific

Synonyms: (E)-N-{[4-Methoxy-3-(prop-2-yn-1-yloxy)phenyl]methylidene}hydroxylamine

043069-1G
101858-994EA 349.6 USD
101858-994
4-Methoxy-3-(2-propynyloxy)benzenecarbaldehydeoxime ≥95%
4-Methoxy-3-(2-propynyloxy)benzenecarbaldehydeoxime

Hazard: Irritant

Formula: C₁₁H₁₁NO₃
MW: 205.22 g/mol
Melting Pt: 118…120 °C
MDL Number: MFCD03001240
CAS Number: 385383-47-1

Order Now

Specification Test Results

Melting point 118-120°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR