[4-Methoxy-3-(2-propynyloxy)phenyl]methanol ≥95%

Supplier: Matrix Scientific

043067-1G
101858-982EA 467.69 USD
101858-982
[4-Methoxy-3-(2-propynyloxy)phenyl]methanol ≥95%
[4-Methoxy-3-(2-propynyloxy)phenyl]methanol

Hazard: Irritant

Formula: C₁₁H₁₂O₃
MW: 192.22 g/mol
Melting Pt: 64…66 °C
MDL Number: MFCD03001238
CAS Number: 385383-49-3

Order Now

Specification Test Results

Melting point 64-66°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR