3-(4-Nitrophenyl)-3-[(2,2,2-trifluoroacetyl)amino]propanoic acid ≥95%

Supplier: Matrix Scientific

043065-500MG
101858-970EA 416.3 USD
101858-970
3-(4-Nitrophenyl)-3-[(2,2,2-trifluoroacetyl)amino]propanoic acid ≥95%
3-(4-Nitrophenyl)-3-[(2,2,2-trifluoroacetyl)amino]propanoic acid

Hazard: Irritant

Formula: C₁₁H₉F₃N₂O₅
Melting Pt: 168…170 °C
MDL Number: MFCD02661624
CAS Number: 439094-19-6

Order Now

Specification Test Results

Melting point 168-170°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR