3-(4-Chlorophenyl)-2-cyclohexyl-1-oxo-1,2,3,4-tetrahydro-4-isoquinolinecarboxylic acid ≥95%

Supplier: Matrix Scientific

043060-500MG 043060-5G
101858-946EA 181.54 USD
101858-946 101858-950
3-(4-Chlorophenyl)-2-cyclohexyl-1-oxo-1,2,3,4-tetrahydro-4-isoquinolinecarboxylic acid ≥95%
3-(4-Chlorophenyl)-2-cyclohexyl-1-oxo-1,2,3,4-tetrahydro-4-isoquinolinecarboxylic acid

Hazard: Irritant

Formula: C₂₂H₂₂ClNO₃
Melting Pt: 197…199 °C
MDL Number: MFCD03001191
CAS Number: 400073-91-8

Order Now

Specification Test Results

Melting point 197-199°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR