2-[(3-Fluorobenzyl)sulfanyl]benzenecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(3-Fluorobenzylthio)benzoic acid

043056-500MG 043056-5G
101858-922EA 375.38 USD
101858-922 101858-926
2-[(3-Fluorobenzyl)sulfanyl]benzenecarboxylic acid ≥95%
2-[(3-Fluorobenzyl)sulfanyl]benzenecarboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₁FO₂S
MW: 262.3 g/mol
Melting Pt: 183…186 °C
MDL Number: MFCD02937499
CAS Number: 385383-44-8

Order Now

Specification Test Results

Melting point 183-186°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR