Methyl-2-[(2,5-dimethylbenzyl)sulfanyl]benzenecarboxylate ≥95%

Supplier: Matrix Scientific

043055-1G
101858-918EA 350.77 USD
101858-918
Methyl-2-[(2,5-dimethylbenzyl)sulfanyl]benzenecarboxylate ≥95%
Methyl-2-[(2,5-dimethylbenzyl)sulfanyl]benzenecarboxylate

Hazard: Irritant

Formula: C₁₇H₁₈O₂S
Melting Pt: 135…137 °C
MDL Number: MFCD03001234
CAS Number: 385383-53-9

Order Now

Specification Test Results

Melting point 135-137°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR