2-[(2,4-Dichlorobenzyl)sulfanyl]benzene carboxylic acid ≥95%

Supplier: Matrix Scientific

043053-1G
101858-906EA 467.69 USD
101858-906
2-[(2,4-Dichlorobenzyl)sulfanyl]benzene carboxylic acid ≥95%
2-[(2,4-Dichlorobenzyl)sulfanyl]benzene carboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₀Cl₂O₂S
MW: 313.2 g/mol
Melting Pt: 217…219 °C
MDL Number: MFCD03001232
CAS Number: 385383-42-6

Order Now

Specification Test Results

Melting point 217-219°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR